Learn More
Med Vet International Sterile Adhesive Surgical Drape w/Povidine, 8" x 12", J1086B, 20/Box
Catalog No. 5000583253
Encompass_Preferred
The majority of human surgeries use clear adhesive drape over the incision site. The rest of the patient is draped with standard non-woven disposable drape material. The clear adhesive drape offers several advantages: Transparency The film is ultra-thin and highly transparent, which gives clear observation to the surgical field for locating anatomical landmarks. Antibacterial Impregnated with Povidine iodine giving the drape a slight orange tint. Very effective against most skin pathogens. Fluid and Pathogen Resistance Sterile barrier prevents fluids or pathogens right up to the edge of the incisior Permeable Allows the skin to breathe and vapor to pass. High Elasticity Follows natural contours for reliable gap-free adherence. Low Sensitivity No irritation to skin or wound. Moderate Viscosity No injury to skin when removing the dressing.Range of sizes, packaged sterile in sealed pouch.
Catalog No. 50-005-83253 Supplier Med Vet International Supplier No. JOR1086B
The majority of human surgeries use clear adhesive drape over the incision site. The rest of the patient is draped with standard non-woven disposable drape material. The clear adhesive drape offers several advantages: Transparency The film is ultra-thin and highly transparent, which gives clear observation to the surgical field for locating anatomical landmarks. Antibacterial Impregnated with Povidine iodine giving the drape a slight orange tint. Very effective against most skin pathogens. Fluid and Pathogen Resistance Sterile barrier prevents fluids or pathogens right up to the edge of the incisior Permeable Allows the skin to breathe and vapor to pass. High Elasticity Follows natural contours for reliable gap-free adherence. Low Sensitivity No irritation to skin or wound. Moderate Viscosity No injury to skin when removing the dressing.Range of sizes, packaged sterile in sealed pouch.