Learn More
BURKLE INC Sampling spatula SteriPlast Bio-PE, sterile, 5.91 in. (150 mm)
Catalog No. 503081388
Encompass_Preferred
The disposable spatula made of green PE is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene and can be completely recycled but is manufactured from renewable raw materials.
As a result of the long, ergonomically-shaped, stable handle and a sharp blade, it is possible to even penetrate directly into containers such as paper or plastic sacks. 
Catalog No. 50-308-1388 Supplier BURKLE INC Supplier No. 53791009
The disposable spatula made of green PE is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene and can be completely recycled but is manufactured from renewable raw materials.
As a result of the long, ergonomically-shaped, stable handle and a sharp blade, it is possible to even penetrate directly into containers such as paper or plastic sacks. 