Learn More
BURKLE INC Sampling spoon SteriPlast Bio-PE, sterile, 0.085 oz. (2,5 ml)
Catalog No. 503081389
Encompass_Preferred
The disposable spoon made of green PE is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The disposable spoon is available in two sizes, 0.08 oz. (2.5 ml) (0.08 oz.) (similar to a teaspoon) and 10 ml (0.34 oz.) (similar to a tablespoon).
Catalog No. 50-308-1389 Supplier BURKLE INC Supplier No. 53791011
The disposable spoon made of green PE is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The disposable spoon is available in two sizes, 0.08 oz. (2.5 ml) (0.08 oz.) (similar to a teaspoon) and 10 ml (0.34 oz.) (similar to a tablespoon).