Learn More
Instech Sterile Catheter Flush Solution, Saline (0.9% NaCl), 50 pcs (10 packs of 5) 1mL in 1mL Syringe, PinPort™ Injector
SDP
Catalog No. NC2302034
Encompass
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains Normal Saline (0.9% NaCl), USP, Sterile Grade. Each 1 mL B-D slip fit luer syringe is filled to 0.5 mL and capped with a sterile PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
AAALAC recommends that pharmaceutical-grade compounds be used in laboratory animal research when available.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
Features
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Presevative-Free
Catalog No. NC2302034 Supplier Instech Supplier No. USPS0.5BD1P50
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains Normal Saline (0.9% NaCl), USP, Sterile Grade. Each 1 mL B-D slip fit luer syringe is filled to 0.5 mL and capped with a sterile PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
AAALAC recommends that pharmaceutical-grade compounds be used in laboratory animal research when available.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
Features
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Presevative-Free