Learn More
Key Scientific Products Sterile Ceramic Beads for CryoCare Organism Preservative System, 50 Vials per Box
SDP
Catalog No. NC9892440
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains sterilized ceramic beads only. Includes 50 vials per box.
Catalog No. NC9892440 Supplier Key Scientific Products Supplier No. KS70G
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains sterilized ceramic beads only. Includes 50 vials per box.