Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superior Glove™ Sterile Cleanroom Cut-Resistant Knit Gloves
Description
- Dyneema™ is lint-free and provides cut and abrasion resistance
- Economy version. 44 grams/pr
- Polyurethane provides secure grip in wet, oily conditions
- Form fitting, breathable, and flexible gloves are ergonomically shaped
- High dexterity and flexibility Washable and bleachable (under 145°C)
- ASTM ANSI Cut Level 2
Specifications
Specifications
| Color | White |
| Product Type | Cut-resistant Gloves |
| Size | 11 |
| Sterility | Sterile |
| For Use With (Application) | Cleanroom environment |
| Glove Cuff Style | Knit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?