Learn More
Electron Microscopy Sciences Cotton-Tipped Applicator, Sterile, 6" OAL
SDP
Catalog No. 50949560
Encompass_Preferred
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length with a 1/12" diameter wooden shaft and head size of 3/16" diameter and length of 5/8". 1/12" diameter wooden shaft and head size is 3/16"(dia) x5/8 "(L).
OAL: 6"
Sterile: Sterile
Catalog No. 50-949-560 Supplier Electron Microscopy Sciences Supplier No. 72310S
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length with a 1/12" diameter wooden shaft and head size of 3/16" diameter and length of 5/8". 1/12" diameter wooden shaft and head size is 3/16"(dia) x5/8 "(L).
OAL: 6"
Sterile: Sterile