Learn More
Med Vet International Sterile Culture Tubes, 5ml, J0556A, 25/bag
Catalog No. 5000582329
Encompass_Preferred
Transparent polystyrene tubes can be used for most routine laboratory procedures. Tubes are supplied with a 2-position ribbed polyethylene cap which can be left open for aerobic work or sealed for anaerobic work. 25/bag.
Catalog No. 50-005-82329 Supplier Med Vet International Supplier No. JOR556A
Transparent polystyrene tubes can be used for most routine laboratory procedures. Tubes are supplied with a 2-position ribbed polyethylene cap which can be left open for aerobic work or sealed for anaerobic work. 25/bag.