Learn More
Alkali Scientific Sterile DI Water 50mL Bottle, 1/Pk
SDP
Catalog No. 5000662828
Encompass_Preferred
Sterile Deionized Water in a 50mL bottle provides a high purity solvent designed for small scale laboratory applications. The compact reduces waste and minimizes contamination risks associated with repeatedly opening larger containers. The water is purified to remove dissolved ions and sterilized to eliminate microbial presence, ensuring reliable performance in sensitive experiments. It is commonly used for reconstituting small volumes of reagents, preparing standards, and serving as a blank or control in analytical procedures. This format is particularly useful in molecular biology, microbiology, and chemical testing where maintaining purity and preventing cross contamination are essential for accurate and reproducible results.
Catalog No. 50-006-62828 Supplier Alkali Scientific Supplier No. SDSB01
Sterile Deionized Water in a 50mL bottle provides a high purity solvent designed for small scale laboratory applications. The compact reduces waste and minimizes contamination risks associated with repeatedly opening larger containers. The water is purified to remove dissolved ions and sterilized to eliminate microbial presence, ensuring reliable performance in sensitive experiments. It is commonly used for reconstituting small volumes of reagents, preparing standards, and serving as a blank or control in analytical procedures. This format is particularly useful in molecular biology, microbiology, and chemical testing where maintaining purity and preventing cross contamination are essential for accurate and reproducible results.