Learn More
Alkali Scientific Sterile DI Water 9.9mL Tube, 10/Pk
SDP
Catalog No. 5000662740
Encompass_Preferred
Sterile Deionized Water in this 9.9mL tube format is specifically designed to simplify microbiological dilution procedures. The pre measured volume allows technicians to create a precise 1:100 dilution by adding 0.1mL of sample, eliminating the need for multiple dilution steps. The water is purified to remove ions and sterilized to prevent contamination, ensuring accurate and reproducible results in laboratory testing. This format improves efficiency by reducing preparation time and minimizing pipetting errors. It is widely used in microbiology, environmental testing, and analytical laboratories for serial dilutions, sample preparation, and procedures requiring consistent dilution accuracy.
Catalog No. 50-006-62740 Supplier Alkali Scientific Supplier No. SDST01
Sterile Deionized Water in this 9.9mL tube format is specifically designed to simplify microbiological dilution procedures. The pre measured volume allows technicians to create a precise 1:100 dilution by adding 0.1mL of sample, eliminating the need for multiple dilution steps. The water is purified to remove ions and sterilized to prevent contamination, ensuring accurate and reproducible results in laboratory testing. This format improves efficiency by reducing preparation time and minimizing pipetting errors. It is widely used in microbiology, environmental testing, and analytical laboratories for serial dilutions, sample preparation, and procedures requiring consistent dilution accuracy.