Learn More
Sigma Aldrich Fine Chemicals Biosciences Sterile disks suitable for
Catalog No. 502901687
Encompass
Can be used to test a variety of antibiotics carbohydrates substrates antiseptics on bacteria in petri dishes. Soak a disc in a SOLUTIONon or apply some SOLUTIONon on the discs. Then let dry and place it in an inoculated agar plate. Each disc will absorb exactly the same amount of liquid.
Catalog No. 50-290-1687 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7414625DISCSF
Can be used to test a variety of antibiotics carbohydrates substrates antiseptics on bacteria in petri dishes. Soak a disc in a SOLUTIONon or apply some SOLUTIONon on the discs. Then let dry and place it in an inoculated agar plate. Each disc will absorb exactly the same amount of liquid.