Learn More
KIMBLE™ Sterile Disposable Glass TD
SDP
Catalog No. 01672127 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Serological Pipet, Single-use
Catalog No. Type
01-672-127 Serological Pipet, Single-use
1 options
NA
Catalog No. 01-672-127 Supplier KIMBLE™ Supplier No. 721001110
Add to Cart
Add to Cart
NA
Color-Coded Serological Pipette Multipack, non-pyrogenic, clean, plugged, and pre-sterilized ideal for precision liquid handling
  • Sterile, plugged short-length pipets perfect for tissue culture work or procedures performed under fume hoods or in confined areas
  • Chemically inert with easy to read blue markings and bidirectional graduations
  • Vented, recloseable multipack for larger volume use permits easy access to several pipets at once without contaminating unused pipets
  • Uniform pulled tip ensures accurate dispensing every time
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Sterility Sterile
Graduated Yes
Color Clear
Volume (Metric) 1 mL
Length (Metric) 274 mm
Type Serological Pipet, Single-use
Disposable Single-use
Fit for Purpose Sterile and PCR Ready
NA