Learn More
Electron Microscopy Sciences Sterile Disposable Pipettes Standard 1 ml, 0.2 ml gradations, 279 mm OAL, Single Wrap package
SDP
Catalog No. 50930576
Encompass_Preferred
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.2 ml
OAL: 279 mm
Package: Single Wrap
Adapter: 4.8 mm
Includes:
Catalog No. 50-930-576 Supplier Electron Microscopy Sciences Supplier No. 709531
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.2 ml
OAL: 279 mm
Package: Single Wrap
Adapter: 4.8 mm
Includes:
Product Title