Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Sterile Disposable Pipettes Standard 5 ml, 0.1 ml gradations, 298 mm OAL, Bulk package
SDP

Supplier:  Electron Microscopy Sciences 709545

Encompass_Preferred

Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.1 ml
OAL: 298 mm
Package: Bulk
Adapter: 8.0 mm
Includes: Suction adapter

Catalog No. 50-930-586


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.