Learn More
Stoelting™ Sterile Disposable Scalpels
SDP
Catalog No. 10000853 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
No. 10
No. 11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-000-853 No. 10
10-000-854 No. 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-853 Supplier Stoelting™ Supplier No. 52186
 Restricted Item
Add to Cart
Add to Cart
Sterile Disposable Scalpels are mounted on plastic handles with fluted finger-grips.
  • Scalpel blade is safely protected by pull-off protector shield.
  • Individually sealed in peel-apart, see-through package.
Specifications
For Use With (Application) Neuroscience
Size No. 10
Material Plastic Handle
Includes Box of 10