Learn More
Electron Microscopy Sciences Sterile Disposable Pipettes Standard 10 ml, 0.1 ml gradations, 302 mm OAL, Single Wrap package
SDP
Catalog No. 50930577
Encompass_Preferred
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.1 ml
OAL: 302 mm
Package: Single Wrap
Adapter: 7.4 mm
Includes:
Catalog No. 50-930-577 Supplier Electron Microscopy Sciences Supplier No. 7095310
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.1 ml
OAL: 302 mm
Package: Single Wrap
Adapter: 7.4 mm
Includes:
Product Title