Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Sterile Disposable Pipettes Standard 50 ml, 0.1 ml gradations, 341 mm OAL, Single Wrap package
SDP

Supplier:  Electron Microscopy Sciences 7095350

Encompass_Preferred

Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to +/-1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Gradations: 0.1 ml
OAL: 341 mm
Package: Single Wrap
Adapter: 8.0 mm
Includes: Suction adapter

Catalog No. 50-930-581


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.