Learn More
Stoelting™ Sterile Disposible Scalpels
SDP
Catalog No. 10000855 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Disposable; Sterile
Catalog No. Model
10-000-855 Disposable; Sterile
1 options
Catalog No. 10-000-855 Supplier Stoelting™ Supplier No. 52188
Add to Cart
Add to Cart
Stoelting™ Sterile Disposable Scaples are mounted on plastic handles with fluted finger-grips.
  • Scalpel blade is safely protected by pull-off protector shield
  • Individually sealed in peel-apart, see-through package.
Specifications
For Use With (Application) Neuroscience
Material Stainless Steel Blades, Plastic Handles
Probe Configuration #15s
Model Disposable; Sterile
Includes Box of 10