Learn More
COPAN Diagnostics Sterile Dry Minitip Foam Swab with Plastic Applicator in Peel Pouch
SDP
Catalog No. 23600984 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Tip Type:
Minitip
Catalog No. Tip Type
23-600-984 Minitip
1 options
Intended to be paired with UTM™ Universal Transport Media, part number 3C047N
Catalog No. 23-600-984 Supplier COPAN Diagnostics Supplier No. 1C061S01
Add to Cart
Add to Cart
Intended to be paired with UTM™ Universal Transport Media, part number 3C047N
Minitip Foam Swab with Plastic Applicator with 80 mm Breakpoint, Sterile
  • 80 mm breakpoint in peel pouch
  • Sterile
Specifications
Shelf Life 3 Years
Sterility Sterile
Handle Material Plastic
Applicator Material Foam
Tip Type Minitip
Tip Material Foam
Breakpoint Yes