Learn More
CELLINK LLC STERILE EMPTY CARTRIDGES 3ML
SDP
Catalog No. NC2456255
Encompass
Empty, sterile cartridges with pistons for bioprinting, mounted with sterile end and tip cap. Packed in Mylar bag.
Catalog No. NC2456255 Supplier CELLINK LLC Supplier No. CSC010311101
Empty, sterile cartridges with pistons for bioprinting, mounted with sterile end and tip cap. Packed in Mylar bag.