Learn More
Mopec Sterile Fiberglass Latent Print Powder Brush
SDP
Catalog No. 22446339 Shop All Mopec Products
Change view
Click to view available options
For Use With (Application):
Latent Fingerprint Development
Catalog No. For Use With (Application)
22-446-339 Latent Fingerprint Development
1 options
Catalog No. 22-446-339 Supplier Mopec Supplier No. FS349
Add to Cart
Add to Cart
Fiberglass bristles apply fingerprint powder while supporting sterile, contamination-free processing. A plastic handle provides controlled use, and individual packaging helps maintain cleanliness and safety while supporting compliance with applicable medical packaging standards.
Construction
  • Transparent packaging
  • Print powder brush
  • Conforms with EN-868-5
  • Sterilization Technique: Ethylene Oxide 15 gr EtO ampoule is used for per 35 dm3 container
Dimensions
  • Handle: 4.13 in. L
  • Brush: 1.77 in. L
Materials
  • Co-polymer film
  • Craft paper
  • Plastic
Specifications
Product Type Latent Print Brush
For Use With (Application) Latent Fingerprint Development