Learn More
Fluotics Agilent Bravo compatible P30, sterile
Catalog No. NC1934096
Encompass_Preferred
Agilent Bravo compatible P30, sterile, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 11484-312
Catalog No. NC1934096 Supplier Fluotics Supplier No. AGI30.ST
Agilent Bravo compatible P30, sterile, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 11484-312