Learn More
Instech Sterile Catheter Flush/Lock solution, Heparin Sodium (10 USP units/mL) in Saline (0.9% NaCl), 1mL in 1mL Syringe, Capped with PNP3M Injector, 50 pcs
SDP
Catalog No. 5000142674
Encompass
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains Heparin Sodium (10 USP units/mL) in Saline (0.9% NaCl) USP, sterile grade. Each 1 cc NORM-JECT-F 1 slip fit luer syringe is filled to 1 mL capped with a PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
AAALAC recommends that pharmaceutical-grade compounds be used in laboratory animal research when available.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring.
For laboratory animal research only. Not approved for human use.
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Sterile Filtered
  • Preservative-Free
Catalog No. 50-001-42674 Supplier Instech Supplier No. USPHS0101NJ1P50
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains Heparin Sodium (10 USP units/mL) in Saline (0.9% NaCl) USP, sterile grade. Each 1 cc NORM-JECT-F 1 slip fit luer syringe is filled to 1 mL capped with a PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
AAALAC recommends that pharmaceutical-grade compounds be used in laboratory animal research when available.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring.
For laboratory animal research only. Not approved for human use.
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Sterile Filtered
  • Preservative-Free