Learn More
Instech Sterile Catheter Flush/Lock Solution, Heparin Sodium (5 USP units/mL) in Saline (0.9% NaCl), 0.5mL in 1mL Syringe, Capped with PNP3M Injector, 50 pcs
SDP
Catalog No. 5000142673
Encompass
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains heparin sodium (5 USP units/mL) in saline (0.9% NaCl) USP, sterile grade. Each 1 cc NORM-JECT-F 1 slip fit luer syringe is filled to 0.5 mL and capped with a PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Preservative-Free
Catalog No. 50-001-42673 Supplier Instech Supplier No. USPHS0050.5NJ1P50
Pharmaceutical-grade catheter flush/lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains heparin sodium (5 USP units/mL) in saline (0.9% NaCl) USP, sterile grade. Each 1 cc NORM-JECT-F 1 slip fit luer syringe is filled to 0.5 mL and capped with a PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Preservative-Free