Learn More
Med Vet International Manuka G 15g Gel Sterile, J1253, 10 Pack
Catalog No. 5000495695
Encompass_Preferred
JorVet sweetens its dressing range. “For over 2,000 years honey has been used as a topical anti-microbial agent and dressing; specifically honey derived from the nectar of the Manuka tree (Leptospermum scoparium) has its own unique chem- ical properties. Manuka honey contains glucose oxidase imparted by the bees during its collection and storage. The enzyme facilitates low level release of hydrogen peroxide when in the presence of wound exudates and thus supports cellular proliferation and metabolism through an antioxidant effect. The high sugar and low pH content creates a high osmotic potential which gives honey excellent debridement and anti-inflammatory properties.One of the biggest difficulties with the use of honey in wound dressings is ensuring that once applied it stays over the wound area. Many attempts to coat dressings have been unsuccessful; often the active honey ingredient is left within the packaging severely reducing the effectiveness of the dressing.
Catalog No. 50-004-95695 Supplier Med Vet International Supplier No. JOR1253
JorVet sweetens its dressing range. “For over 2,000 years honey has been used as a topical anti-microbial agent and dressing; specifically honey derived from the nectar of the Manuka tree (Leptospermum scoparium) has its own unique chem- ical properties. Manuka honey contains glucose oxidase imparted by the bees during its collection and storage. The enzyme facilitates low level release of hydrogen peroxide when in the presence of wound exudates and thus supports cellular proliferation and metabolism through an antioxidant effect. The high sugar and low pH content creates a high osmotic potential which gives honey excellent debridement and anti-inflammatory properties.One of the biggest difficulties with the use of honey in wound dressings is ensuring that once applied it stays over the wound area. Many attempts to coat dressings have been unsuccessful; often the active honey ingredient is left within the packaging severely reducing the effectiveness of the dressing.