Learn More
Rockland Immunochemicals Sterile Human Plasma In Sodium Heparin 50 mL
SDP
Catalog No. 501943969
Encompass_Preferred
Human Plasma is used as a supplement to cell culture media. Human Plasma provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation.
Catalog No. 50-194-3969 Supplier Rockland Immunochemicals Supplier No. D519060050
Human Plasma is used as a supplement to cell culture media. Human Plasma provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation.