Learn More
Med Vet International Sterile IV Administration Set, 10dr/mL, 100in with Injection Site, Luer Slip
Catalog No. NC0196893
Encompass_Preferred
The Oasis IV administration set features a universal kink-resistant tubing with smooth action roller clamp, and Y injection site. Available in 10, 15, 20 and 60 drop per mL Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 100'
Catalog No. NC0196893 Supplier Med Vet International Supplier No. IV10
The Oasis IV administration set features a universal kink-resistant tubing with smooth action roller clamp, and Y injection site. Available in 10, 15, 20 and 60 drop per mL Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 100'