Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Irradiated NeutraKlean Solution
BIOPHARMAGREENER_CHOICE

Catalog No. 19200021 Shop All Contec Inc. Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-200-021 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-200-021 Supplier Contec™ Supplier No. SBT103NK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for the low level cleaning required after maintenance and to remove disinfectant residues and general soil.

  • Neutral pH after irradiation and suitable for use in all areas of cleanroom as gentle on surfaces
  • Free from contamination and particulates, filtered to 0.22μm under Grade A airflow in Grade C cleanroom
  • Low foaming detergent action
  • Gamma irradiated
  • Low level of surfactant
  • Trigger spray can be set to jet or spray
  • Trigger spray and bag in bottle protected system
  • Double bagged packed in linear tear packaging

Specifications

Product Type NeutraKlean Detergent
pH 6.5 to 7.5
Sterility Sterile
Composition 0.3% amine oxide in purified water (EP)
Description Sterile by gamma radiation; Trigger spray with jet or spray option
Filtered 0.22μm
For Use With (Application) For cleaning of life science cleanrooms
Includes Certificates of analysis
Quantity 6/Cs.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.