Learn More
Sklar Instruments Econo Sterile Operating Scissors, 5 1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel, CS/50.
SDP
Catalog No. 501109786 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Sterile Operating Scissors, 5 1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel, CS/50. Econo Sterile Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches. They are sold in cases of 50.
Catalog No. 50-110-9786 Supplier Sklar Instruments Supplier No. 962520A
Econo Sterile Operating Scissors, 5 1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel, CS/50. Econo Sterile Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches. They are sold in cases of 50.