Learn More
Biotang Inc Sterile PBS, 500 ml: 10 mM Sodium phosphate dibasic, 2 mM potassium phosphate monobasic, 2.7 mM potassium chloride, 137 mM sodium chloride, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water. 0.22-micron filtered and/or autoclaved. BM-220S
Catalog No. 50155853 Shop All BIOTANG Inc Products
Encompass
Sterile PBS, 500 ml: 10 mM Sodium phosphate dibasic, 2 mM potassium phosphate monobasic, 2.7 mM potassium chloride, 137 mM sodium chloride, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water. 0.22-micron filtered and/or autoclaved. BM-220S.
Catalog No. 50-155-853 Supplier Biotang Inc Supplier No. BTBB531
Sterile PBS, 500 ml: 10 mM Sodium phosphate dibasic, 2 mM potassium phosphate monobasic, 2.7 mM potassium chloride, 137 mM sodium chloride, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water. 0.22-micron filtered and/or autoclaved. BM-220S.