Learn More
VAI Sterile Pens
Catalog No. 19546585 Shop All VAI Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
19-546-585 Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-546-585 Supplier VAI Supplier No. VAIPEN01
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Sterile Pens are gamma irradiated sterile blue ink pens compatible with Core2Write Cleanroom Documentation forms, logbooks, tags, and labels and CleanPrint 10 synthetic writing substrate.
  • Abrasion, tear resistant, and smear resistant
  • Lightweight and flexible but exceptionally durable
  • Cellulose free and extremely low in particulates and shedding features
  • Triple Bagged
Specifications
Color Blue
Sterility Sterile
Thickness 1.0 mm