Learn More
Cole-Parmer Thermo Scientific Nalgene 2019-2000 Sterile PETG Media Bottles, 2000 mL; 12/cs
Catalog No. NC1677217 Shop All Cole-Parmer Products
Encompass
Durable PETG (Polyethylene Terephthalate G Copolymer) bottles with HDPE closures are radiation-sterilized and nonpyrogenic—eliminates washing, depyrogenation, and autoclaving steps. Reduced permeability to CO2/O2 allows long-term media storage. Tamper-resistant band around closure and neck of each bottle ensures sterility. Bottles come in shrink-wrapped trays.
Catalog No. NC1677217 Supplier Cole-Parmer Supplier No. 0625460
Durable PETG (Polyethylene Terephthalate G Copolymer) bottles with HDPE closures are radiation-sterilized and nonpyrogenic—eliminates washing, depyrogenation, and autoclaving steps. Reduced permeability to CO2/O2 allows long-term media storage. Tamper-resistant band around closure and neck of each bottle ensures sterility. Bottles come in shrink-wrapped trays.