Learn More
Electron Microscopy Sciences Sterile Petri Dish 100x20 mm, 100 ml, 90x20 mm
SDP
Catalog No. 501900268
Encompass_Preferred
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. With three venting ribs into the underside of the lid to prevent condensation build-up.
Volume: 100 ml
Size: 90 x 20 mm
Sterile: Sterile
Catalog No. 50-190-0268 Supplier Electron Microscopy Sciences Supplier No. 7067820
Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by "corona effect". Made of polystyrene. With three venting ribs into the underside of the lid to prevent condensation build-up.
Volume: 100 ml
Size: 90 x 20 mm
Sterile: Sterile