Learn More
StatLab Sterile Phosphate Buffer

Description
- Ready-to-use single bottle of sterilized buffer solution
- No weighing, mixing, or autoclave time is needed for this ready-to-use sterilized buffer
- Compatible with multiple automated TB systems
- 100 mL bottle
Specifications
Specifications
| Form | Solution |
| Manufacturing Origin | United States |
| Product Type | Phosphate Buffer |
| Sterility | Sterile |
| Target Organism Class | Mycobacteria |
| Test Type | Qualitative QC |
| For Use With (Application) | Compatible with automated TB systems |
| Volume (Metric) Specimen | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.