Learn More
Andwin Scientific STERILE PIPETTE 500/PK
SDP
Supplier:  Andwin Scientific 709628S
Encompass_Preferred
The graduated transfer pipets are specifically made to be utilized for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, and several other applications. This product has a measured length of 15.5 centimeters and a stem diameter of 4.6 millimeters. The sterile pipets are packed individually in a bulk quantity of 500 pieces.
Catalog No. 50-930-611