Learn More
Thomson Instrument Company Well Plate Lid, Sterile | Use w/ All 96- & 24-Well Plates
SDP
Catalog No. 501489311
Encompass_Preferred
Thomson Instrument Company Well Plate lid fits the entire offering of 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Lid is clear and designed to work with SLAS specifications. Many Thomson well plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. 50-148-9311 Supplier Thomson Instrument Company Supplier No. 981948
Thomson Instrument Company Well Plate lid fits the entire offering of 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Lid is clear and designed to work with SLAS specifications. Many Thomson well plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.