Learn More
STEMCELL Technologies CellAdhere™ Collagen I-Coated Flask, 25 cm², Size: 5 Units
SDP
Supplier:  STEMCELL Technologies 1000348
Encompass
Culture attachment-dependent cells and tissues in collagen I-coated flasks from CellAdhereᵀᴹ. Non-pyrogenic and made with clear polystyrene for superior optical clarity, these vent-capped flasks are coated with high-purity collagen I, derived from rat tail tendon for optimal cell attachment. The canted neck of these flasks offers easy pipette and cell scraper access to the corners. Flasks are available in packs of 5 in 25 cm², 75 cm², and 175 cm² volumes, or individually wrapped 225 cm² volume.
Catalog No. 50-204-5518