Learn More
Cardinal Health™ Sterile Powder Free Vinyl Gloves
Catalog No. 191500728 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
19-150-0728 Medium
1 options
This item is not returnable. View return policy
Catalog No. 19-150-0728 Supplier Cardinal Health™ Supplier No. 2D7012PF
This item is not returnable. View return policy
Smooth finish glove with excellent grip capabilities
  • Glove fits closer to the hand and wrist — better than traditional PVC gloves
  • Better puncture resistance than most PVC gloves
  • Demonstrated permeation resistance to a number of commonly used hospital and lab chemicals
Specifications
Hand Ambidextrous
Material Vinyl
Powdered Powder Free
Size Medium
Sterility Sterile