Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Sterile Ringer’s Solutions, Frog, Innovating Science™
SCI_ED

Catalog No. S37743 Shop All Aldon Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37743 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37743 Supplier Innovating Science™ Supplier No. IS5066
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solution for frogs

Specifications

Chemical Name or Material Sodium chloride
Color Colorless
CAS Mixture, 7647-14-5, 144-55-8, 7447-40-7, 10035-04-8
Boiling Point ∼100°C (water)
Melting Point ∼0°C (water)
Solubility Information Complete in water
Grade Reagent
Packaging Plastic bottle
Vapor Pressure 14 (water)
Vapor Density 0.7 (water)
Quantity 200 mL
Physical Form Liquid
Show More Show Less

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.