Learn More
BURKLE INC Sampling scoop SteriPlast Bio-PE, sterile, 1.69 oz. (50 ml)
Catalog No. 503081240
Encompass_Preferred
Sampling scoop SteriPlast, sterile, made from sustainable green PE bio-plastic. The innovative Bio-PE material combines the proven properties of conventional polyethylene — such as durability, chemical resistance, and food safety — with an eco-friendly production from renewable raw materials. Fully recyclable for minimal environmental impact. Sterile packaging ensures hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity:1.69 oz. (50 ml).
Catalog No. 50-308-1240 Supplier BURKLE INC Supplier No. 53791003
Sampling scoop SteriPlast, sterile, made from sustainable green PE bio-plastic. The innovative Bio-PE material combines the proven properties of conventional polyethylene — such as durability, chemical resistance, and food safety — with an eco-friendly production from renewable raw materials. Fully recyclable for minimal environmental impact. Sterile packaging ensures hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity:1.69 oz. (50 ml).