Learn More
BURKLE INC Sampling scoop SteriPlast Bio-PE, sterile, 5.07 oz. (150 ml)
Catalog No. 503081387
Encompass_Preferred
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
Catalog No. 50-308-1387 Supplier BURKLE INC Supplier No. 53791007
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.