Learn More
BURKLE INC Scoop SteriPlast, PS, sterile, 16.91 oz. (500 ml), with cover
Supplier:  BURKLE INC 53781016
Encompass_Preferred
Sterile SteriPlast disposable sampling scoops, made from high-quality white polystyrene, are produced in Class 7 (ISO) cleanroom conditions, assembled, individually packaged, and gamma-sterilized for maximum cleanliness. Designed for sensitive sampling of powders, granules, and pastes, they ensure safe, contamination-free handling. The ergonomic shape with a slightly raised front edge allows stable placement without sample loss. As single-use tools, they require no cleaning or sterilization, saving time and resources while minimizing staff exposure to hazardous materials. Individually packaged, they remain sterile until use, eliminating cross-contamination risks between different samples or products. Ideal for laboratory, pharmaceutical, and food industry applications requiring strict hygiene and reliable sample integrity.
Catalog No. 50-308-1278