Learn More
Research Products International Corp Sterile Sampler Scoops, 2 Ounce Capacity, 100 per Case
SDP
Catalog No. 50998981
Encompass
Sterileware scoops are the perfect shape and size for taking samples, especially from wide-mouth containers. Designed to hold various capacities, these one-piece sterile scoops are molded of high-impact polystyrene plastic.
2oz sampling scoops have a long handle that increases the distance between the hand and sample
When placed on a flat surface they remain upright; can be used as weighing boats
Made from FDA grade materials
Individually sealed and sterilized in a polyethylene bag ready for one time use and disposal
Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
This item is not returnable. View return policy
Catalog No. 50-998-981 Supplier Research Products International Corp Supplier No. H36902
Sterileware scoops are the perfect shape and size for taking samples, especially from wide-mouth containers. Designed to hold various capacities, these one-piece sterile scoops are molded of high-impact polystyrene plastic.
2oz sampling scoops have a long handle that increases the distance between the hand and sample
When placed on a flat surface they remain upright; can be used as weighing boats
Made from FDA grade materials
Individually sealed and sterilized in a polyethylene bag ready for one time use and disposal
Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.