Learn More
Cole-Parmer Scienceware F37944-0000 Sterile Polystyrene (PS) Serrated Square Tip Sampling Tongs, 4.25" L; 25/cs
Catalog No. NC0161463 Shop All Cole-Parmer Products
Encompass
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals. Individually packaged, sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped wih a Certificate of Processing for Sterilization.
Catalog No. NC0161463 Supplier Cole-Parmer Supplier No. 0644302
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals. Individually packaged, sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped wih a Certificate of Processing for Sterilization.