Learn More
Roboz Surgical STERILE SCALPEL BLADE NO10 BOX
SDP
Supplier:  Roboz Surgical RS980110
Encompass
Sterile Scalpel Blades #10; Interchangeable; For Use With Handles #3 & #7; Box Of 100
Catalog No. NC9182948