Learn More
Marketlab, Inc. Sterile Serological Pipettes
Description
- Fits all major brands of pipettors
- Aerosol barrier plug prevents both liquid and aerosols from contaminating the pipettor
- Calibration is certified for accuracy and meets E934
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Sterile |
| Color | Clear |
| Volume (Metric) | 25 mL |
| Type | Serological Pipet, Single-use |
| Disposable | Single-use |