Learn More
VAI Sterile Sharpie Markers
Catalog No. 19546586 Shop All VAI Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
19-546-586 Black
1 options
Catalog No. 19-546-586 Supplier VAI Supplier No. VAISHA01
Add to Cart
Add to Cart
Sterile Sharpie Markers are gamma irradiated sterile sharpie markers are compatible with Core2Write Cleanroom Documentation forms, logbooks, tags, and labels and CleanPrint 10 synthetic writing substrate.
  • Permanent Black Ink
  • Fine Tip
  • Abrasion, tear resistant, and smear resistant
  • Lightweight and flexible but exceptionally durable
  • Cellulose free and extremely low in particulates and shedding features
  • Individually Double Bagged
Specifications
Product Type Permanent Marker
Color Black
Sterility Sterile