Learn More
Kuhner Shaker Inc STERILE SINGLE-USE BAG
Supplier:  Kuhner Shaker Inc NSMX760003
Encompass
Pack of 3 Special Perfusion-Ready Sterile Single-Use Bags for use with Kuhner Shaker's SB10-X Orbitally Shaken Bioreactors. Includes bio-weldable tubing (with additional ports for perfusion processes) and integrated (non-invasive) pH and DO Optical Sensors.
Catalog No. 50-257-0322