Learn More
Kuhner Shaker Inc STERILE SINGLE-USE BAG
Catalog No. 502570323
Encompass
Pack of 3 Standard Sterile Single-Use Bags for use with Kuhner Shaker's SB10-X Orbitally Shaken Bioreactors. Includes bio-weldable tubing and integrated (non-invasive) pH and DO Optical Sensors. (H)
Catalog No. 50-257-0323 Supplier Kuhner Shaker Inc Supplier No. NSMX760001
Pack of 3 Standard Sterile Single-Use Bags for use with Kuhner Shaker's SB10-X Orbitally Shaken Bioreactors. Includes bio-weldable tubing and integrated (non-invasive) pH and DO Optical Sensors. (H)