Learn More
Kuhner Shaker Inc STERILE SINGLE-USE BAG
Supplier:  Kuhner Shaker Inc NSM7500053
Encompass
Pack of 3 Standard Sterile Single-Use Bags for use with Kuhner Shaker's SB50-X Orbitally Shaken Bioreactors. Includes bio-weldable tubing and integrated (non-invasive) pH and DO Optical Sensors. (H)
Catalog No. 50-257-0327
Add to Cart
Request a Quote